Learn More
Lectrodryer DUAL SCALE DIAL THERMOMETER
SDP
Catalog No. NC1923538
Encompass_Preferred
2" DIAL, 4 1/2" STEM, -100*C-50*C, -150*F-120*F, STAINLESS STEEL
Catalog No. NC1923538 Supplier Lectrodryer Supplier No. D53701
2" DIAL, 4 1/2" STEM, -100*C-50*C, -150*F-120*F, STAINLESS STEEL