Description
Roller adjustment permits fast, easy measuring. Polished-steel, dual scale vernier measures inner diameter, outer diameter, and depth. Range is 0–120mm in 1mm divisions, and 0–5 in. in 1/16 in. divisions. This instrument is readable on a vernier scale to 0.1mm and 1/128 in..
Specifications
Specifications
| For Use With (Equipment) | Dual scale roller |
| Material | Steel |
| For Use With (Application) | Ideal to measure inner diameter, outer diameter and depth |
| Product Type | Adjustment Caliper |