Learn More
Fisherbrand™ Dual Solution Reservoir
Catalog No. 14127652 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Natural
Catalog No. Color
14-127-652 Natural
1 options
Catalog No. 14-127-652
Add to Cart
Add to Cart
Reservoir shape helps guide and protect pipette tips during use.
  • Multi-functional reagent reservoir has a single compartment 50 mL basin on one side and 12 slotted 5 mL basins on the other side
  • Angled wells guide pipette tips toward a center position to help avoid cross contamination and waste
  • Individual 5 mL basin wells numerical imprinted to identify samples
  • Cover included to prevent contamination
Specifications
Color Natural
Capacity (Metric) 5/50 mL
Dimensions (L x W x H) 4.9 x 2 x 1.5 in.
Material Polypropylene
Sterility Non-sterile
Type Multichannel Reservoir
Autoclavable Autoclavable
Channels 12
Disposable Reusable
Bottom Shape Convoluted