Learn More
YSI 3P LifeScience 2747 Glucose/Lactate Standard, 1.80 g/L Glucose 0.45 g/L Lactate
Catalog No. 50200144
Encompass
Glucose and Lactate standard solution. Calibrator solution for the YSI 2300 Stat Plus. May also be used as calibrator or check solution with the YSI 2700 Analyzer.
Catalog No. 50-200-144 Supplier YSI 3P LifeScience Supplier No. 2747
Glucose and Lactate standard solution. Calibrator solution for the YSI 2300 Stat Plus. May also be used as calibrator or check solution with the YSI 2700 Analyzer.