Learn More
Stoelting™ Dual Syringe Infusion Pump
SDP
Catalog No. 10000866 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Infusion
Catalog No. Model
10-000-866 Infusion
1 options
Catalog No. 10-000-866 Supplier Stoelting™ Supplier No. 53200
Stoelting™ Dual Syringe Infusion Pump is ideal for delivering accurate and precise amounts of fluids for a multitude of syringe pump applications
  • Two-syringe infusion pump combines a broad speed range
  • Holds a wide range of syringe sizes to meet the requirements of virtually any laboratory application.
  • Includes infusing calibrant into a mass spectrometer or reaction chamber, long term drug infusions to animals and general infusion applications.
Specifications
For Use With (Application) Neuroscience
Model Infusion