Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kuhner Shaker Inc DUAL TABLE 85 MM FOR 2 E/F TAB

Catalog No. 502569964
Encompass

Dual Table 85 mm for 2 E/F Tablare ISF1-X, ISF4-X, SS-X

Catalog No. 50-256-9964 Supplier Kuhner Shaker Inc Supplier No. SMX651303
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dual Table 85 mm for 2 E/F Tablare ISF1-X, ISF4-X, SS-X

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.