Learn More
Stoelting™ Dual Temperature Cautery Kit
SDP
Catalog No. 10001021 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Battery-operated
Catalog No. Model
10-001-021 Battery-operated
1 options
Catalog No. 10-001-021 Supplier Stoelting™ Supplier No. 59017
Add to Cart
Add to Cart
Easily replaceable, autoclavable, nickel-chrome tips require less power and provide longer battery life. The convenient battery spacer converts the handpiece from high temperature to low temperature.
Specifications
For Use With (Application) Neuroscience
Temperature (English) Range 2200°F and 1300°F
Model Battery-operated