Learn More
Fisherbrand™ Autoclavable Biohazard Waste Bags
SDPSCI_ED
Catalog No. S35910 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
19 x 23 in.
Catalog No. Dimensions (L x W)
S35910 19 x 23 in.
1 options
This item is not returnable. View return policy
Catalog No. S35910
This item is not returnable. View return policy
Dual tested compliant
  • Integral steam processing indicator that turns brown when autoclaved
  • Bags comply with US DOT 49CFR Sections 173-197 and specific state regulations
  • Operating temperature tolerance up to 285degreeFdegreeAvailable in various sizes
  • Bag with black print
  • Gauge: 2mil
Specifications
Color Red
Dimensions (L x W) 19 x 23 in.