Learn More
Electron Microscopy Sciences Dual-Tip Cryo-Markers™, Red
SDP
Catalog No. 501930722
Encompass_Preferred
High quality cryogenic permanent ink markers with durable, heavy duty, long-lasting nibs. These 2-in-1 markers have dual tips: fine (0.7 mm) and extra-fine (0.3 mm) and are easy to handle for applications in bio-medical laboratories. They are permanent and water-resistant (all colors equal). Excellent performance on plastic, glass, metal, cloth and other surfaces.
Color: Red
Catalog No. 50-193-0722 Supplier Electron Microscopy Sciences Supplier No. 6205501R
High quality cryogenic permanent ink markers with durable, heavy duty, long-lasting nibs. These 2-in-1 markers have dual tips: fine (0.7 mm) and extra-fine (0.3 mm) and are easy to handle for applications in bio-medical laboratories. They are permanent and water-resistant (all colors equal). Excellent performance on plastic, glass, metal, cloth and other surfaces.
Color: Red