Learn More
World Precision Instrument Dual Tool-Holder Micromanipulator-MD4R
SDP
Catalog No. 502538722
Encompass
A small and compact micromanipulator for manual manipulation in all three axes (x, y and z), the MD4 is equipped with a mounting bracket for a second tool or elec­trode holder which can be positioned in the x and y axes in­de­pen­dent of the manipulator and may also be tilted and swiveled by two fine-adjust screws. Scales allow readings of coarse adjustment with an ac­cu­ra­cy of 100 µm. Additional x-axis fine control is achieved with a micrometer screw with a resolution of 10 µm.
Catalog No. 50-253-8722 Supplier World Precision Instrument Supplier No. MD4R
A small and compact micromanipulator for manual manipulation in all three axes (x, y and z), the MD4 is equipped with a mounting bracket for a second tool or elec­trode holder which can be positioned in the x and y axes in­de­pen­dent of the manipulator and may also be tilted and swiveled by two fine-adjust screws. Scales allow readings of coarse adjustment with an ac­cu­ra­cy of 100 µm. Additional x-axis fine control is achieved with a micrometer screw with a resolution of 10 µm.