Learn More
Kuhner Shaker Inc DUAL TRAY F180 WITH 2 X F TRAY
Catalog No. 502570008
Encompass
Dual Tray F180 with 2 x F trays (800 x 420mm) set 180mm apart...Plain trays (without pre-drilled holes). Top tray removable. Shaking speed limited: 150 rpm at 50mm orbit diameter, 250 rpm at 25mm.
Catalog No. 50-257-0008 Supplier Kuhner Shaker Inc Supplier No. SMX3016
Dual Tray F180 with 2 x F trays (800 x 420mm) set 180mm apart...Plain trays (without pre-drilled holes). Top tray removable. Shaking speed limited: 150 rpm at 50mm orbit diameter, 250 rpm at 25mm.