Learn More
Kuhner Shaker Inc DUAL TRAY FU180 WITH 2 X FU TR
Catalog No. 502570009
Encompass
Dual Tray FU180 with 2 x FU trays (800 x 420mm) set 180mm apart...Universal trays with pre-drilled holes. Top tray removable. Shaking speed limited: 150 rpm at 50mm orbit diameter, 250 rpm at 25mm.
Catalog No. 50-257-0009 Supplier Kuhner Shaker Inc Supplier No. SMX3015
Dual Tray FU180 with 2 x FU trays (800 x 420mm) set 180mm apart...Universal trays with pre-drilled holes. Top tray removable. Shaking speed limited: 150 rpm at 50mm orbit diameter, 250 rpm at 25mm.