Learn More
Kuhner Shaker Inc DUAL TRAY FU200 WITH 2 X FU TR
Catalog No. 502570007
Encompass
Dual Tray FU200 with 2 x FU trays (800 x 420mm) set 200mm apart...Universal trays with pre-drilled holes. Top tray removable. Shaking speed limited: 150 rpm at 50mm orbit diameter, 250 rpm at 25mm.
Catalog No. 50-257-0007 Supplier Kuhner Shaker Inc Supplier No. SMX3017
Dual Tray FU200 with 2 x FU trays (800 x 420mm) set 200mm apart...Universal trays with pre-drilled holes. Top tray removable. Shaking speed limited: 150 rpm at 50mm orbit diameter, 250 rpm at 25mm.