Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Instech Two channel tether kit for rats. 12in spring tether, SIP22/4 injection port on striped line, 375/22PS swivel on clear line, 24in COEX tubing and luer stub. Sterile. Connects to VAHD115AB rat harnesses.
SDP

Catalog No. 5081116
Encompass

The Instech KVAHD115T is a complete tether kit designed for two-channel Vascular Access Harness™ (VAH™) systems in rats. It enables continuous access to one fluid channel through a swivel while providing a side port for intermittent manual access to a second channel. This sterile kit maintains a closed system to ensure catheter patency and animal safety during specialized research like bile sampling or simultaneous infusion and sampling.

Features

  • Application: Continuous infusion and intermittent sampling for two-channel rat VAH™
  • Compatibility: Specifically for use with the VAHD115AB two-channel rat harness
  • Dimensions: 12in (30cm) spring tether with 24in (60cm) CoEx™ extension tubing
  • Components: VAHD115T tether, SIP22/4 injection port, 375/22P swivel, and LS22 luer stub

Catalog No. 50-811-16 Supplier Instech Supplier No. KVAHD115T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Instech KVAHD115T is a complete tether kit designed for two-channel Vascular Access Harness™ (VAH™) systems in rats. It enables continuous access to one fluid channel through a swivel while providing a side port for intermittent manual access to a second channel. This sterile kit maintains a closed system to ensure catheter patency and animal safety during specialized research like bile sampling or simultaneous infusion and sampling.

Features

  • Application: Continuous infusion and intermittent sampling for two-channel rat VAH™
  • Compatibility: Specifically for use with the VAHD115AB two-channel rat harness
  • Dimensions: 12in (30cm) spring tether with 24in (60cm) CoEx™ extension tubing
  • Components: VAHD115T tether, SIP22/4 injection port, 375/22P swivel, and LS22 luer stub

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.