Learn More
Electron Microscopy Sciences Write-On™ Dual Core Write On Tape Dispenser
SDP
Catalog No. 5029790
Encompass_Preferred
This tape dispenser offers you a convenient writing platform, suitable for both left- and right-handed users. The epoxy-coated platform with finger holes and built-in cutting edge, make dispensing and cutting the tape a simple one-handed action. The weighted base with non-skid bottom pads assures stability. The 3" (76 mm) core fits standard tape rolls up to 1" (25 mm) wide. Weight: 4 lbs (1.8kg). The dispenser accepts series 77010-tapes shown above. The tape is not included. Holds 1 & 3" core Write-On Label Tape, standard base. Measures: 302 x 143 x 160 mm H.
Catalog No. 50-297-90 Supplier Electron Microscopy Sciences Supplier No. 7700120
This tape dispenser offers you a convenient writing platform, suitable for both left- and right-handed users. The epoxy-coated platform with finger holes and built-in cutting edge, make dispensing and cutting the tape a simple one-handed action. The weighted base with non-skid bottom pads assures stability. The 3" (76 mm) core fits standard tape rolls up to 1" (25 mm) wide. Weight: 4 lbs (1.8kg). The dispenser accepts series 77010-tapes shown above. The tape is not included. Holds 1 & 3" core Write-On Label Tape, standard base. Measures: 302 x 143 x 160 mm H.