Learn More
MilliporeSigma DUALEX TO UNIT
Catalog No. NC9535209
Encompass
Transducer Protector, Sterile, Diameter: 28.6 mm, Filtration Area: 3.9 sq. cm, Filter Pore Size: 0.22 micrometer, Max. Pressure: 1.7 bar, Wettability: Hydrophobic, Fitting Inlet: Female Luer slip, Fitting Outlet: Male Luer slip, 50/PK
Catalog No. NC9535209 Supplier MilliporeSigma Supplier No. SLGVS25XS
Add to Cart
Add to Cart
Transducer Protector, Sterile, Diameter: 28.6 mm, Filtration Area: 3.9 sq. cm, Filter Pore Size: 0.22 micrometer, Max. Pressure: 1.7 bar, Wettability: Hydrophobic, Fitting Inlet: Female Luer slip, Fitting Outlet: Male Luer slip, 50/PK