Learn More
MilliporeSigma DUALEX ULTRA TP PK/50
Catalog No. NC9606632
Encompass
Dualex Transducer Protector, 50/Pk.;28.6mm dia,Filtration Area: 3.9cm2,Filter Pore Size: 0.22micrometer, Max Operating Temperature: 45degreeC; Max Pressure: 1.7bar;Filter Diameter: 25mm,Device Color: clear inlet/blue outlet; Height: 25mm
This item is not returnable. View return policy
Catalog No. NC9606632 Supplier MilliporeSigma Supplier No. SLGVS25US
Dualex Transducer Protector, 50/Pk.;28.6mm dia,Filtration Area: 3.9cm2,Filter Pore Size: 0.22micrometer, Max Operating Temperature: 45degreeC; Max Pressure: 1.7bar;Filter Diameter: 25mm,Device Color: clear inlet/blue outlet; Height: 25mm