Learn More
Electron Microscopy Sciences Dual Size SEM Pin Stub Holder, 15 mm
SDP
Catalog No. 501930553
Encompass_Preferred
A simple holder which will hold ⅛" (3.1 mm) pin stubs on one side, or any other solid stub in the other side. Useful for specimen preparation, coating, freeze drying, as well as storage.
Diameter: 15 mm
Catalog No. 50-193-0553 Supplier Electron Microscopy Sciences Supplier No. EMS003
A simple holder which will hold ⅛" (3.1 mm) pin stubs on one side, or any other solid stub in the other side. Useful for specimen preparation, coating, freeze drying, as well as storage.
Diameter: 15 mm