Learn More
Thermo Scientific™ DualSpin Rotor Body
Catalog No. 13101503 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
accuSpin™ 8C Benchtop Centrifuge
Catalog No. For Use With (Equipment)
13-101-503 accuSpin™ 8C Benchtop Centrifuge
1 options
Catalog No. 13-101-503 Supplier Thermo Scientific™ Supplier No. 75008813
  • $891.00
    $819.65 / Each of 1
    Save $71.35
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Run common clinical IVD blood tubes, gel tubes, 10 mL standard syringes and 15 mL conical tubes with the Thermo Scientific™ DualSpin™ rotor.
TRUSTED_SUSTAINABILITY
Specifications
Material Lightweight Composite
Type Rotor Body
For Use With (Equipment) accuSpin™ 8C Benchtop Centrifuge