Learn More
Medchemexpress LLC HY-50737A 5mg Medchemexpress, DUB-IN-2 CAS:924296-19-5 Purity:>98%
SDP
Catalog No. 502030950
Encompass_Preferred
Medchemexpress, HY-50737A 5mg DUB-IN-2 CAS:924296-19-5 DUB-IN-2 is a potent deubiquitinase inhibitor with an IC50 of 0.28 μM for USP8. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0950 Supplier Medchemexpress LLC Supplier No. HY50737A5MG
Medchemexpress, HY-50737A 5mg DUB-IN-2 CAS:924296-19-5 DUB-IN-2 is a potent deubiquitinase inhibitor with an IC50 of 0.28 μM for USP8. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.