Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DUB-IN-3 | 924296-17-3 | 99.0% | C16H9N5O | 100 MG
SDP

Catalog No. 5000358742
Encompass_Preferred

DUB-IN-3 (compound 22c) is a potent and selective inhibitor of the deubiquitinating enzyme USP8 with an IC50 value of 0.56 μM. This compound is a solid, light yellow to yellow in appearance, with a purity of 99.0%. It holds promise for research in various fields including cancer, neurodegenerative diseases, inflammation, viral infection, and cardiovascular disease.

  • Potent and selective inhibitor of USP8
  • Suitable for research into cancer
  • Aids research in neurodegenerative diseases
  • Useful for inflammation studies
  • Applicable for viral infection research
  • Supports cardiovascular disease research
  • Purity of 99.0%
  • Chemical formula: C16H9N5O
  • Soluble in DMSO (25 mg/mL)

Catalog No. 50-003-58742 Supplier Medchemexpress LLC Supplier No. HY50737100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DUB-IN-3 (compound 22c) is a potent and selective inhibitor of the deubiquitinating enzyme USP8 with an IC50 value of 0.56 μM. This compound is a solid, light yellow to yellow in appearance, with a purity of 99.0%. It holds promise for research in various fields including cancer, neurodegenerative diseases, inflammation, viral infection, and cardiovascular disease.

  • Potent and selective inhibitor of USP8
  • Suitable for research into cancer
  • Aids research in neurodegenerative diseases
  • Useful for inflammation studies
  • Applicable for viral infection research
  • Supports cardiovascular disease research
  • Purity of 99.0%
  • Chemical formula: C16H9N5O
  • Soluble in DMSO (25 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.