Learn More
Medchemexpress LLC Dub-in-3 | 924296-17-3 | 99.0% | C16H9N5O | 5 MG
SDP
Catalog No. 5000357906
Encompass_Preferred
DUB-IN-3 is a potent and selective inhibitor of the deubiquitinating enzyme USP8 with an IC50 value of 0.56 μM. It shows promise for research in cancer, neurodegenerative diseases, inflammation, viral infection, and cardiovascular disease. This compound also affects the viability of HCT116 colon and PC-3 prostate cancer cell lines with IC50 values in the 0.5-1.5 μM range.
  • Potent and selective USP8 inhibitor
  • Potential research applications in cancer
  • Potential research applications in neurodegenerative diseases
  • Potential research applications in inflammation
  • Potential research applications in viral infections
  • Potential research applications in cardiovascular disease
  • Affects viability of HCT116 colon and PC-3 prostate cancer cell lines
Catalog No. 50-003-57906 Supplier Medchemexpress LLC Supplier No. HY507375MG
DUB-IN-3 is a potent and selective inhibitor of the deubiquitinating enzyme USP8 with an IC50 value of 0.56 μM. It shows promise for research in cancer, neurodegenerative diseases, inflammation, viral infection, and cardiovascular disease. This compound also affects the viability of HCT116 colon and PC-3 prostate cancer cell lines with IC50 values in the 0.5-1.5 μM range.
  • Potent and selective USP8 inhibitor
  • Potential research applications in cancer
  • Potential research applications in neurodegenerative diseases
  • Potential research applications in inflammation
  • Potential research applications in viral infections
  • Potential research applications in cardiovascular disease
  • Affects viability of HCT116 colon and PC-3 prostate cancer cell lines