Learn More
Medchemexpress LLC HY-12963 5mg Medchemexpress, Dubermatinib CAS:1341200-45-0 Purity:>98%
SDP
Catalog No. 502028933
Encompass_Preferred
Medchemexpress, HY-12963 5mg Dubermatinib CAS:1341200-45-0 Dubermatinib (TP-0903) is a potent and selective Axl receptor tyrosine kinase inhibitor with an IC50 value of 27 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8933 Supplier Medchemexpress LLC Supplier No. HY129635MG
Medchemexpress, HY-12963 5mg Dubermatinib CAS:1341200-45-0 Dubermatinib (TP-0903) is a potent and selective Axl receptor tyrosine kinase inhibitor with an IC50 value of 27 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.