Learn More
Enterprise Technology Solutions Duck Brand Carpet Tape, Indoor/Outdoor, White
SDP
Catalog No. 5000830454
Encompass_Preferred
Indoor/Outdoor Carpet Tape features an incredibly strong adhesive for permanent carpet installation. Fiberglass-reinforced design increases its overall durability to provide a lasting hold on surfaces. Moisture-resistant construction provides adheres reliably to surfaces whether it's secured indoors or outdoors. Carpet tape is perfect for installing, holding, seaming or repairing carpet seams.
Catalog No. 50-008-30454 Supplier Enterprise Technology Solutions Supplier No. DUC286372
Indoor/Outdoor Carpet Tape features an incredibly strong adhesive for permanent carpet installation. Fiberglass-reinforced design increases its overall durability to provide a lasting hold on surfaces. Moisture-resistant construction provides adheres reliably to surfaces whether it's secured indoors or outdoors. Carpet tape is perfect for installing, holding, seaming or repairing carpet seams.