Learn More
Enterprise Technology Solutions Duck Brand Packaging Tape,HP 235,f/Recycled Boxes,48mmx100m,6/PK,CL
SDP
Catalog No. 5000830215
Encompass_Preferred
Hot-melt packaging tape for recycled cartons features an enhanced adhesive system that offers higher shear, tack and adhesion to produce better seals - no matter the recycled content of the carton. Special-release coating allows HP 235 Packaging Tape to consistently unwind off the roll with low force for efficient tape consumption. Strong adhesive maintains a secure hold to endure the rigors of shipping and storage. Packaging tape is perfect for use on medium-weight cartons in everyday manufacturing and shipping environments.
Catalog No. 50-008-30215 Supplier Enterprise Technology Solutions Supplier No. DUC242763
Hot-melt packaging tape for recycled cartons features an enhanced adhesive system that offers higher shear, tack and adhesion to produce better seals - no matter the recycled content of the carton. Special-release coating allows HP 235 Packaging Tape to consistently unwind off the roll with low force for efficient tape consumption. Strong adhesive maintains a secure hold to endure the rigors of shipping and storage. Packaging tape is perfect for use on medium-weight cartons in everyday manufacturing and shipping environments.