Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Ducks Unlimited™ Ear Muffs

Catalog No. 19155305 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black, Gray
Pink
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-155-305 Pink
19-155-304 Black, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-155-305 Supplier Pyramex™ Supplier No. DUPM8010P

Comfortable over-the-head design with a convenient fold-away feature for easy storage.

  • Low-Profile Design
  • Soft Foam Ear Cups
  • Folding Headband
  • Individually Packaged

Specifications

Color Pink
ANSI S3.1974 Yes
Cushion Material Foam
Cushion Color Black
For Use With (Application) Construction, Energy and Utilities, General Manufacturing, Mining, Oil and Gas, Transportation
Model DUPM8010P
Packaging Type Poly bag
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB
Headband Color Black
Style Over-the-head
Foldable Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.