Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Ducks Unlimited™ Hearing and Safety Eyewear Combo Kit

Catalog No. 19155302 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Includes:
Ear Muffs and Clear Shooting Glasses
Ear Muffs and Orange Shooting Glasses
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-155-302 Ear Muffs and Clear Shooting Glasses
19-155-303 Ear Muffs and Orange Shooting Glasses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-155-302 Supplier Pyramex™ Supplier No. DUCOMBO5710

Combo Kit includes one pair of PM8010 Ear Muffs along with one pair of Venture™ 3 Safety Glasses.

  • Convenient Combo Kit includes one pair of ear muffs and one pair of safety glasses
  • Venture™ Eyewear features soft, adjustable, ventilated nose piece
  • Eyewear frame is ventilated to prevent fogging
  • Lens is scratch resistant
  • Ear Muff features low-profile design with soft ear cups
  • Fold-away padded headband for comfortability and easy storage

Specifications

Color Black, Gray
ANSI S3.1974 Yes
Cushion Material Foam
Cushion Color Black
For Use With (Application) Variety of industrial and recreational applications.
Model DUCOMBO5710
Pairs per Package 1 Ea. (Ear Muff/Safety Eyewear)
Noise Reduction Rating (NRR) 26 dB
Product Type Combo Kit
Headband Color Black
Style Over-the-head
Includes Ear Muffs and Clear Shooting Glasses
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.