Learn More
Pyramex™ Ducks Unlimited™ Safety Eyewear
Catalog No. 19155306 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Green
Orange
Vermillion
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-306 Clear
19-155-307 Vermillion
19-155-308 Green
19-155-309 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-155-306 Supplier Pyramex™ Supplier No. DUSB6610D
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Shooting safety glasses
  • Soft PVC nose piece generates a cushion for a comfort fit
  • Fashionable stream line design. 9.5 base curve lens
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Frame Color Black
    Lens Material Polycarbonate