Learn More
MSC Duct Incorporated Duct Hose Adapter, 4 in Galvanized, 1 ea
Catalog No. 5000503863
Encompass
This product is a galvanized duct hose adapter designed for use in various ducting systems. It provides a secure connection for 4-inch diameter hoses.
  • Inside diameter: 4 in
  • Overall length: 4 in
  • Material: galvanized
  • Manufactured in the USA
  • Hose adapter type fitting
Catalog No. 50-005-03863 Supplier MSC Supplier No. 03233541
This product is a galvanized duct hose adapter designed for use in various ducting systems. It provides a secure connection for 4-inch diameter hoses.
  • Inside diameter: 4 in
  • Overall length: 4 in
  • Material: galvanized
  • Manufactured in the USA
  • Hose adapter type fitting