Learn More
MSC Duct Incorporated Galvanized Reducer, 10x6 in, 20 Gage, 1 ea
Catalog No. 5000503864
Encompass
This galvanized reducer duct fitting is designed to connect ductwork of different diameters, ensuring efficient airflow transition. It features a durable construction suitable for industrial ventilation systems.
  • Constructed from galvanized material for corrosion resistance
  • Provides a 10-6 in inside diameter reduction
  • Overall length of 10 in for versatile installation
  • Features 20 gage thickness for robust performance
Catalog No. 50-005-03864 Supplier MSC Supplier No. 03233715
This galvanized reducer duct fitting is designed to connect ductwork of different diameters, ensuring efficient airflow transition. It features a durable construction suitable for industrial ventilation systems.
  • Constructed from galvanized material for corrosion resistance
  • Provides a 10-6 in inside diameter reduction
  • Overall length of 10 in for versatile installation
  • Features 20 gage thickness for robust performance