Learn More
Grainger NSI Industries Duct Seal, Chemically Inert, Nontoxic, 1 lb, 1 ea
Catalog No. 502773798
Encompass_Preferred
This duct sealing compound is designed for sealing around electrical service entries, junction boxes, and other devices where weather-proofing is critical. The 1 lb package provides a versatile, non-toxic sealing material that adheres to most clean, dry surfaces.
  • Will stay soft and pliable for several years and has an excellent shelf life.
  • It is chemically inert, inhibiting no chemical activity and is non-toxic.
  • Contains no asbestos or mineral fibers and no mineral oil which can shorten the effective life of the product.
  • Will not damage wire covers, glass, wood, painted surfaces, plastics or other materials it may contact.
  • Remains pliable through a wide range of temperatures and will adhere to most clean dry surfaces.
Catalog No. 50-277-3798 Supplier Grainger Supplier No. 173T06
This duct sealing compound is designed for sealing around electrical service entries, junction boxes, and other devices where weather-proofing is critical. The 1 lb package provides a versatile, non-toxic sealing material that adheres to most clean, dry surfaces.
  • Will stay soft and pliable for several years and has an excellent shelf life.
  • It is chemically inert, inhibiting no chemical activity and is non-toxic.
  • Contains no asbestos or mineral fibers and no mineral oil which can shorten the effective life of the product.
  • Will not damage wire covers, glass, wood, painted surfaces, plastics or other materials it may contact.
  • Remains pliable through a wide range of temperatures and will adhere to most clean dry surfaces.