Learn More
Grainger Edwards Signaling SuperDuct Four-Wire Duct Smoke Detector, Photoelectric, 1 ea
Catalog No. 502766945
Encompass_Preferred
The SuperDuct™ SD-CJ is a four-wire photoelectric duct smoke detector designed to monitor airflow and identify the presence of smoke within HVAC systems. This device triggers alarm signaling systems to alert building occupants of fire hazards and offers integrated environmental compensation to ensure performance.
  • Features patented differential sensing for environmental compensation
  • Performs automatic sensitivity adjustments every eight minutes
  • Includes a dedicated dirty and trouble LED indicator on each unit
  • Supports RJ-45 interconnection capability
  • Enables remote testing via key or magnet without requiring detector cover removal
  • Complies with CSFM, UL, and ULC standards
Catalog No. 50-276-6945 Supplier Grainger Supplier No. 16X427
The SuperDuct™ SD-CJ is a four-wire photoelectric duct smoke detector designed to monitor airflow and identify the presence of smoke within HVAC systems. This device triggers alarm signaling systems to alert building occupants of fire hazards and offers integrated environmental compensation to ensure performance.
  • Features patented differential sensing for environmental compensation
  • Performs automatic sensitivity adjustments every eight minutes
  • Includes a dedicated dirty and trouble LED indicator on each unit
  • Supports RJ-45 interconnection capability
  • Enables remote testing via key or magnet without requiring detector cover removal
  • Complies with CSFM, UL, and ULC standards