Learn More
Grainger Dura-Blok Duct Support Block, Recycled Rubber, 1000 lbs. Capacity
Catalog No. 502755170
Encompass_Preferred
This duct support block provides a versatile and durable solution for rooftop support applications including pipe, HVAC, duct, conduit, and cable trays. Designed for structural and environmental integrity, it eliminates the need for roof penetrations, helping to maximize roof life. The system is manufactured from 100% recycled rubber and features electro-plated steel hardware for corrosion resistance.
  • Dampens vibration to maintain environmental integrity.
  • Features a 1 in. (25.4 mm) gap between blocks to allow water to flow freely.
  • Includes a reflective strip on both sides for added visibility.
  • Ultimate load rating of 1,000 lbs. (4.45 kN).
  • Resistant to UV, freezing, and thawing.
  • Qualifies for LEED credits.
Catalog No. 50-275-5170 Supplier Grainger Supplier No. 15E540
This duct support block provides a versatile and durable solution for rooftop support applications including pipe, HVAC, duct, conduit, and cable trays. Designed for structural and environmental integrity, it eliminates the need for roof penetrations, helping to maximize roof life. The system is manufactured from 100% recycled rubber and features electro-plated steel hardware for corrosion resistance.
  • Dampens vibration to maintain environmental integrity.
  • Features a 1 in. (25.4 mm) gap between blocks to allow water to flow freely.
  • Includes a reflective strip on both sides for added visibility.
  • Ultimate load rating of 1,000 lbs. (4.45 kN).
  • Resistant to UV, freezing, and thawing.
  • Qualifies for LEED credits.