Learn More
MSC DUCT TAPE
Catalog No. 5000803211
Encompass
tesa® 64663 is a duct tape engineered with an aggressive adhesive, suitable for demanding industrial applications such as heavy-duty draping and securing. It features a high-grade cloth backing, providing excellent tensile properties while maintaining tearability for ease of application. The tape's high coating weight makes it suitable for use on very rough surfaces.
  • Aggressive adhesive for demanding industrial applications
  • High-grade cloth backing for excellent tensile properties
  • Tearable for easy application
  • High coating weight for use on very rough surfaces
  • Suitable for industrial, commercial, utility, and automotive applications
  • Effective for sealing, fastening, fixing, closing, packaging, bundling, holding, seaming, protecting, wrapping, and repairing
Catalog No. 50-008-03211 Supplier MSC Supplier No. 41928292
tesa® 64663 is a duct tape engineered with an aggressive adhesive, suitable for demanding industrial applications such as heavy-duty draping and securing. It features a high-grade cloth backing, providing excellent tensile properties while maintaining tearability for ease of application. The tape's high coating weight makes it suitable for use on very rough surfaces.
  • Aggressive adhesive for demanding industrial applications
  • High-grade cloth backing for excellent tensile properties
  • Tearable for easy application
  • High coating weight for use on very rough surfaces
  • Suitable for industrial, commercial, utility, and automotive applications
  • Effective for sealing, fastening, fixing, closing, packaging, bundling, holding, seaming, protecting, wrapping, and repairing