Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Value Collection Duct Tape, 2 in, 60 yd, 1 Roll

Catalog No. 501919246
Encompass

This duct tape is designed for various applications, including bundling, light repair, and sealing. It features a rubber adhesive for strong adhesion.

  • Width: 2 in
  • Length: 60 yd
  • Thickness: 10 mil
  • Color: Beige
  • Adhesive material: Rubber
  • Applications: Bundling, light repair, and sealing

Catalog No. 50-191-9246 Supplier MSC Supplier No. 39588595
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This duct tape is designed for various applications, including bundling, light repair, and sealing. It features a rubber adhesive for strong adhesion.

  • Width: 2 in
  • Length: 60 yd
  • Thickness: 10 mil
  • Color: Beige
  • Adhesive material: Rubber
  • Applications: Bundling, light repair, and sealing

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.