Learn More
MSC Tape Logic Duct Tape, 10 mil, Brown, 16 rolls
Catalog No. 501918607
Encompass
This all-purpose duct tape provides a tough, moisture-resistant adhesive solution for various applications. Featuring an aggressive natural rubber adhesive, it adheres to a wide variety of surfaces for sealing, bundling, and repair tasks.
  • Extremely versatile for various applications
  • Multi-layer construction for durability
  • Conformable and moisture resistant
  • Tear by hand for easy application
  • Aggressive rubber adhesive adheres to multiple surfaces
  • Long-lasting performance for indoor and outdoor use
  • Service temperature range: 32–140 °F
  • Tensile strength: 31 lb/in
  • Total thickness: 10 mil
  • Dimensions: 3 in x 60 yd
Catalog No. 50-191-8607 Supplier MSC Supplier No. 39581970
This all-purpose duct tape provides a tough, moisture-resistant adhesive solution for various applications. Featuring an aggressive natural rubber adhesive, it adheres to a wide variety of surfaces for sealing, bundling, and repair tasks.
  • Extremely versatile for various applications
  • Multi-layer construction for durability
  • Conformable and moisture resistant
  • Tear by hand for easy application
  • Aggressive rubber adhesive adheres to multiple surfaces
  • Long-lasting performance for indoor and outdoor use
  • Service temperature range: 32–140 °F
  • Tensile strength: 31 lb/in
  • Total thickness: 10 mil
  • Dimensions: 3 in x 60 yd