Learn More
Grainger Nashua Professional Grade Duct Tape, 11 Mil, 12/cs.
Catalog No. 502756168
Encompass_Preferred
Professional grade 11 mil duct tape engineered for demanding HVAC and industrial applications. This single-coated tape features a polyethylene-coated cloth backing paired with an aggressive rubber-based adhesive to ensure a secure, permanent bond across a variety of surfaces, including irregular substrates.
  • Excellent balance of tensile strength and tearability
  • Aggressive adhesion to rough surfaces
  • Conforms well to irregular surfaces
  • Tears straight and does not curl
  • Low VOC content
  • Rated for service temperatures from 40°F to 200°F
  • Tested in accordance with UL 723 and ASTM E84-91A
Catalog No. 50-275-6168 Supplier Grainger Supplier No. 15R465
Professional grade 11 mil duct tape engineered for demanding HVAC and industrial applications. This single-coated tape features a polyethylene-coated cloth backing paired with an aggressive rubber-based adhesive to ensure a secure, permanent bond across a variety of surfaces, including irregular substrates.
  • Excellent balance of tensile strength and tearability
  • Aggressive adhesion to rough surfaces
  • Conforms well to irregular surfaces
  • Tears straight and does not curl
  • Low VOC content
  • Rated for service temperatures from 40°F to 200°F
  • Tested in accordance with UL 723 and ASTM E84-91A