Learn More
Grainger Nashua Professional Grade Duct Tape, Black, 11 mil, 2-3/4 in. x 60 yd.
Catalog No. 502753811
Encompass_Preferred
This 11 mil professional grade duct tape is designed for demanding HVAC, construction, and manufacturing applications. Featuring a high-quality polyethylene-coated cloth backing and a reliable rubber adhesive, this tape provides strong adhesion and conforms well to irregular surfaces for effective sealing and repair.
  • Adheres to 80 oz./in. and provides a tensile strength of 26 lb./in.
  • Service temperature range of 40°F to 200°F.
  • Tears straight and resists curling for efficient application.
  • Low VOC content meets UL 723 standards and can contribute toward LEED EQ Credit 4.1.
  • Suitable for indoor and outdoor maintenance, duct sealing, and bundling.
Catalog No. 50-275-3811 Supplier Grainger Supplier No. 15R442
This 11 mil professional grade duct tape is designed for demanding HVAC, construction, and manufacturing applications. Featuring a high-quality polyethylene-coated cloth backing and a reliable rubber adhesive, this tape provides strong adhesion and conforms well to irregular surfaces for effective sealing and repair.
  • Adheres to 80 oz./in. and provides a tensile strength of 26 lb./in.
  • Service temperature range of 40°F to 200°F.
  • Tears straight and resists curling for efficient application.
  • Low VOC content meets UL 723 standards and can contribute toward LEED EQ Credit 4.1.
  • Suitable for indoor and outdoor maintenance, duct sealing, and bundling.