Learn More
Grainger 3M Extra Heavy Duty Duct Tape, Polyethylene Film, 60 yd., 24/Case
Catalog No. 502756805
Encompass_Preferred
This industrial strength, heavy-duty duct tape features an abrasion-resistant polyethylene film over a dense, moisture-resistant cloth scrim backing. Engineered with an aggressive rubber adhesive, it provides a strong, instant bond to most surfaces for demanding applications such as maintenance, repair, construction, and HVAC. It is designed to offer high conformability to irregular surfaces and provides a tight seal.
  • Features a rubber adhesive that adheres aggressively to most surfaces
  • Constructed with a moisture-resistant, polyethylene-coated cloth scrim
  • Offers industrial strength, abrasion resistance, and easy-tear properties
  • Supports temperatures from 0°F to +200°F (+93°C)
  • Meets ASTM D 3759, ASTM E 84, and UL 723 standards
  • Thickness of 10.0 mils (0.25 mm) with a tensile strength of 34 lbs./in. (595 N/100 mm)
Catalog No. 50-275-6805 Supplier Grainger Supplier No. 15F767
This industrial strength, heavy-duty duct tape features an abrasion-resistant polyethylene film over a dense, moisture-resistant cloth scrim backing. Engineered with an aggressive rubber adhesive, it provides a strong, instant bond to most surfaces for demanding applications such as maintenance, repair, construction, and HVAC. It is designed to offer high conformability to irregular surfaces and provides a tight seal.
  • Features a rubber adhesive that adheres aggressively to most surfaces
  • Constructed with a moisture-resistant, polyethylene-coated cloth scrim
  • Offers industrial strength, abrasion resistance, and easy-tear properties
  • Supports temperatures from 0°F to +200°F (+93°C)
  • Meets ASTM D 3759, ASTM E 84, and UL 723 standards
  • Thickness of 10.0 mils (0.25 mm) with a tensile strength of 34 lbs./in. (595 N/100 mm)