Learn More
Grainger 3M Performance Plus Duct Tape, Polyethylene Film, 2 in, 60 yd, Slate Blue
Catalog No. 502754152
Encompass_Preferred
Performance Plus Duct Tape is a high-strength adhesive tape featuring a polyethylene film laminated to a cloth scrim with a rubber adhesive. Designed for versatility, this tape provides UV, water, and abrasion resistance while conforming to irregular surfaces. It is suitable for diverse indoor and outdoor applications including bundling, sealing, splicing, reinforcing, and temporary repair.
  • Removable with little or no adhesive residue for up to six months after application
  • Water-resistant backing provides a moisture-resistant seal
  • Tested for flame spread using UL 723 and ASTM E84 methods
  • Withstands temperatures up to 93°C (200°F) for short durations
  • Hand-tearable for easy, on-site application
  • Sticks immediately and remains secure for up to one year
  • Conforms to irregular surfaces without lifting
Catalog No. 50-275-4152 Supplier Grainger Supplier No. 15F807
Performance Plus Duct Tape is a high-strength adhesive tape featuring a polyethylene film laminated to a cloth scrim with a rubber adhesive. Designed for versatility, this tape provides UV, water, and abrasion resistance while conforming to irregular surfaces. It is suitable for diverse indoor and outdoor applications including bundling, sealing, splicing, reinforcing, and temporary repair.
  • Removable with little or no adhesive residue for up to six months after application
  • Water-resistant backing provides a moisture-resistant seal
  • Tested for flame spread using UL 723 and ASTM E84 methods
  • Withstands temperatures up to 93°C (200°F) for short durations
  • Hand-tearable for easy, on-site application
  • Sticks immediately and remains secure for up to one year
  • Conforms to irregular surfaces without lifting