Learn More
Grainger Nashua Premium Duct Tape, PE-Coated Cloth, 13 mil, 1 7/8 in x 60 yd, 24/Cs.
Catalog No. 502756126
Encompass_Preferred
This premium 13 mil heavy duty duct tape consists of a natural rubber adhesive on a polyethylene-coated cloth backing. Engineered for high performance in industrial, mining, and entertainment applications, this tape offers robust durability and reliable adhesion across various surfaces. It is designed to resist moisture, solvents, chemicals, abrasion, and punctures, providing a dependable solution for demanding bundling, sealing, and temporary securing tasks.
  • Features high tensile strength for difficult applications.
  • Resists curl, delamination, and tearing.
  • Offers a high-tack adhesive that bonds to smooth and rough textures.
  • Features hand-tearable construction for a clean, straight tear.
  • Operates in temperatures from 4°C to 93°C (40°F to 200°F).
  • Meets RoHS standards and contributes to LEED® credits for low-emitting materials.
Catalog No. 50-275-6126 Supplier Grainger Supplier No. 15R449
This premium 13 mil heavy duty duct tape consists of a natural rubber adhesive on a polyethylene-coated cloth backing. Engineered for high performance in industrial, mining, and entertainment applications, this tape offers robust durability and reliable adhesion across various surfaces. It is designed to resist moisture, solvents, chemicals, abrasion, and punctures, providing a dependable solution for demanding bundling, sealing, and temporary securing tasks.
  • Features high tensile strength for difficult applications.
  • Resists curl, delamination, and tearing.
  • Offers a high-tack adhesive that bonds to smooth and rough textures.
  • Features hand-tearable construction for a clean, straight tear.
  • Operates in temperatures from 4°C to 93°C (40°F to 200°F).
  • Meets RoHS standards and contributes to LEED® credits for low-emitting materials.