Learn More
Grainger Nashua 357 Duct and Repair Tape, 13 mil, PE-Coated Cloth, 60 yd
Catalog No. 502757075
Encompass_Preferred
This premium 13 mil duct tape is designed for heavy-duty HVAC, construction, and maintenance applications. Featuring a durable PE-coated cloth backing and a thick natural rubber adhesive, this tape provides superior strength and adheres effectively to rough or dirty surfaces, both indoors and outdoors.
  • High tensile strength of 50 lb/in.
  • Moisture-, chemical-, and abrasion-resistant.
  • Operating temperature range of 40°F to 200°F.
  • Conforms to curves and corners without delaminating.
  • Straight-tear and curl-resistant properties for ease of use.
  • UL 723 and UL listed for performance safety.
  • Suitable for bundling, sealing, seaming, and general repairs.
Catalog No. 50-275-7075 Supplier Grainger Supplier No. 15R446
This premium 13 mil duct tape is designed for heavy-duty HVAC, construction, and maintenance applications. Featuring a durable PE-coated cloth backing and a thick natural rubber adhesive, this tape provides superior strength and adheres effectively to rough or dirty surfaces, both indoors and outdoors.
  • High tensile strength of 50 lb/in.
  • Moisture-, chemical-, and abrasion-resistant.
  • Operating temperature range of 40°F to 200°F.
  • Conforms to curves and corners without delaminating.
  • Straight-tear and curl-resistant properties for ease of use.
  • UL 723 and UL listed for performance safety.
  • Suitable for bundling, sealing, seaming, and general repairs.