Learn More
Grainger 3M Vinyl Duct Tape 3903, Green, 2 in, 50 yd
Catalog No. 502754002
Encompass_Preferred
3M™ Vinyl Duct Tape 3903 is a versatile, hand-tearable, general purpose tape designed for a wide range of industrial applications. Featuring an embossed vinyl backing paired with a rubber adhesive, this tape provides excellent conformability to irregular surfaces and moisture resistance, making it suitable for temporary repairs, identification, bundling, and protective sealing.
  • Embossed vinyl backing resists abrasion and many chemicals while sealing out dust, dirt, and moisture.
  • Hand-tearable construction allows for easy application without curling.
  • Effective across a temperature range of 40°F to 200°F.
  • Suitable for indoor use in assembly, manufacturing, and general maintenance environments.
  • Ideal for color coding, process tagging, and enhancing decor.
Catalog No. 50-275-4002 Supplier Grainger Supplier No. 15F773
3M™ Vinyl Duct Tape 3903 is a versatile, hand-tearable, general purpose tape designed for a wide range of industrial applications. Featuring an embossed vinyl backing paired with a rubber adhesive, this tape provides excellent conformability to irregular surfaces and moisture resistance, making it suitable for temporary repairs, identification, bundling, and protective sealing.
  • Embossed vinyl backing resists abrasion and many chemicals while sealing out dust, dirt, and moisture.
  • Hand-tearable construction allows for easy application without curling.
  • Effective across a temperature range of 40°F to 200°F.
  • Suitable for indoor use in assembly, manufacturing, and general maintenance environments.
  • Ideal for color coding, process tagging, and enhancing decor.