Learn More
Grainger Nashua Premium Grade Duct Tape, Olive Drab, 13 mil, 4 in. x 60 yd.
Catalog No. 502756189
Encompass_Preferred
This heavy duty, premium grade duct tape is designed for demanding maintenance, repair, and construction applications. Featuring a polyethylene-coated cloth backing with a high-performance natural rubber adhesive, this tape provides excellent durability, moisture resistance, and chemical protection. It is engineered to conform to curves and corners while maintaining high tensile strength and a clean, straight tear.
  • Adhesion to steel: 95 oz./in.
  • Tensile strength: 50 lb/in.
  • Service temperature: 40°F to 200°F
  • Tested in accordance with UL723
  • Low VOC content
  • Moisture-, abrasion-, and chemical-resistant
Catalog No. 50-275-6189 Supplier Grainger Supplier No. 15R451
This heavy duty, premium grade duct tape is designed for demanding maintenance, repair, and construction applications. Featuring a polyethylene-coated cloth backing with a high-performance natural rubber adhesive, this tape provides excellent durability, moisture resistance, and chemical protection. It is engineered to conform to curves and corners while maintaining high tensile strength and a clean, straight tear.
  • Adhesion to steel: 95 oz./in.
  • Tensile strength: 50 lb/in.
  • Service temperature: 40°F to 200°F
  • Tested in accordance with UL723
  • Low VOC content
  • Moisture-, abrasion-, and chemical-resistant