Learn More
Grainger Polyken Duct Tape, Medium Duty, Red, 12.0 mil, 48 mm x 55 M
Catalog No. 502757154
Encompass_Preferred
Polyken 226 is a medium-duty, single-coated pressure sensitive tape designed for demanding industrial applications. It features a robust PE-coated cloth backing and an aggressive natural rubber-based adhesive, providing excellent bonding performance on diverse substrates including steel, austenitic stainless steel, and nickel-based alloys. This tape is suitable for critical environments such as power plants and ships.
  • Features a PE-coated cloth backing for high tensile strength
  • Utilizes an aggressive rubber adhesive for reliable bonding
  • Tested in accordance with UL 723
  • Operates within a temperature range of 4°C to 93°C (40°F to 200°F)
  • Meets ASME NQA-1 and NRC Reg 1.38 specifications
  • Provides excellent moisture and solvent resistance
Catalog No. 50-275-7154 Supplier Grainger Supplier No. 15R472
Polyken 226 is a medium-duty, single-coated pressure sensitive tape designed for demanding industrial applications. It features a robust PE-coated cloth backing and an aggressive natural rubber-based adhesive, providing excellent bonding performance on diverse substrates including steel, austenitic stainless steel, and nickel-based alloys. This tape is suitable for critical environments such as power plants and ships.
  • Features a PE-coated cloth backing for high tensile strength
  • Utilizes an aggressive rubber adhesive for reliable bonding
  • Tested in accordance with UL 723
  • Operates within a temperature range of 4°C to 93°C (40°F to 200°F)
  • Meets ASME NQA-1 and NRC Reg 1.38 specifications
  • Provides excellent moisture and solvent resistance