Learn More
Grainger Nashua Duct Tape, PE-Coated Cloth, Red, 11 mil, 60 yd
Catalog No. 502756371
Encompass_Preferred
This 11 mil professional grade duct tape features a PE-coated cloth backing and a natural rubber adhesive, providing an excellent balance of tensile strength and tearability. It is designed for aggressive adhesion to rough surfaces and offers superior holding power for various industrial and maintenance applications.
  • Features a PE-coated cloth backing for strength and durability
  • Natural rubber adhesive ensures aggressive adhesion to rough surfaces
  • Low VOC content can contribute toward satisfying EQ Credit 4.1
  • Suitable for duct sealing, HVAC contractor uses, and maintenance tasks
  • Provides reliable performance for bundling, color coding, and general repairs
Catalog No. 50-275-6371 Supplier Grainger Supplier No. 15R439
This 11 mil professional grade duct tape features a PE-coated cloth backing and a natural rubber adhesive, providing an excellent balance of tensile strength and tearability. It is designed for aggressive adhesion to rough surfaces and offers superior holding power for various industrial and maintenance applications.
  • Features a PE-coated cloth backing for strength and durability
  • Natural rubber adhesive ensures aggressive adhesion to rough surfaces
  • Low VOC content can contribute toward satisfying EQ Credit 4.1
  • Suitable for duct sealing, HVAC contractor uses, and maintenance tasks
  • Provides reliable performance for bundling, color coding, and general repairs