Learn More
Grainger 3M Vinyl Duct Tape, Red, 2 in. x 50 yd., 6.5 mil
Catalog No. 502754003
Encompass_Preferred
3M™ Vinyl Duct Tape 3903 is a versatile, hand-tearable, general-purpose tape featuring a reliable rubber adhesive and a durable, embossed vinyl backing. It is designed for diverse indoor applications including temporary repairs, identification, color-coding, bundling, and protecting surfaces in assembly, manufacturing, and warehouse settings.
  • Rubber adhesive provides fast, secure bonding to irregular surfaces
  • Embossed vinyl backing seals against dust, dirt, and moisture while resisting abrasion and corrosion
  • Hand-tearable construction allows for easy application without curling
  • Suitable for performance temperatures ranging from 40°F to 200°F
  • Ideal for process tagging, marking, highlighting hazards, and general maintenance
  • Meets ASTM standards D3330, D3652, and D3759
Catalog No. 50-275-4003 Supplier Grainger Supplier No. 15F774
3M™ Vinyl Duct Tape 3903 is a versatile, hand-tearable, general-purpose tape featuring a reliable rubber adhesive and a durable, embossed vinyl backing. It is designed for diverse indoor applications including temporary repairs, identification, color-coding, bundling, and protecting surfaces in assembly, manufacturing, and warehouse settings.
  • Rubber adhesive provides fast, secure bonding to irregular surfaces
  • Embossed vinyl backing seals against dust, dirt, and moisture while resisting abrasion and corrosion
  • Hand-tearable construction allows for easy application without curling
  • Suitable for performance temperatures ranging from 40°F to 200°F
  • Ideal for process tagging, marking, highlighting hazards, and general maintenance
  • Meets ASTM standards D3330, D3652, and D3759