Learn More
Grainger Polyken 226 Duct Tape, PE-Coated Cloth, 12 Mil, 60 yd
Catalog No. 502756129
Encompass_Preferred
This premium nuclear grade duct tape is a high-performance, single-coated tape designed for demanding industrial applications. With a 12 mil thickness and a polyethylene-coated cloth backing, it provides exceptional durability, conformability, and aggressive adhesion to various surfaces. It is engineered to meet stringent nuclear, industrial, and safety standards, making it suitable for use in power plants, shipbuilding, and general maintenance environments.
  • Meets ASME NQA-1 and NRC Reg 1.38 regulations
  • Features moisture, solvent, and abrasion resistance
  • Provides crisp, straight tears and high conformability
  • Suitable for bonding with nickel-based alloys and austenitic stainless steels
  • Low VOC content and contributes to LEED EQ Credit 4.1
  • Suitable for indoor and outdoor applications
  • Adhesion to steel of 50 oz/in
Catalog No. 50-275-6129 Supplier Grainger Supplier No. 15R470
This premium nuclear grade duct tape is a high-performance, single-coated tape designed for demanding industrial applications. With a 12 mil thickness and a polyethylene-coated cloth backing, it provides exceptional durability, conformability, and aggressive adhesion to various surfaces. It is engineered to meet stringent nuclear, industrial, and safety standards, making it suitable for use in power plants, shipbuilding, and general maintenance environments.
  • Meets ASME NQA-1 and NRC Reg 1.38 regulations
  • Features moisture, solvent, and abrasion resistance
  • Provides crisp, straight tears and high conformability
  • Suitable for bonding with nickel-based alloys and austenitic stainless steels
  • Low VOC content and contributes to LEED EQ Credit 4.1
  • Suitable for indoor and outdoor applications
  • Adhesion to steel of 50 oz/in