Learn More
Grainger 3M Duct Tape, Polyethylene Coated Cloth, Silver, 24/Pk.
Catalog No. 502759574
Encompass_Preferred
3M™ Duct Tape 3939 is a high-performance tape designed for demanding industrial and construction applications. It features a waterproof polyethylene film laminated to a cloth scrim backing with an aggressive rubber adhesive, providing excellent holding power, moisture resistance, and conformability for a variety of tasks including sealing, patching, bundling, and reinforcing.
  • Features a rubber adhesive that adheres aggressively to most surfaces for a tight seal.
  • Polyethylene-coated cloth scrim resists moisture to prolong bond integrity.
  • Offers high conformability and ease of tearing by hand.
  • Low tack roll edges help maintain cleaner surfaces and reduce waste.
  • Suitable for temperatures up to 93°C (200°F).
  • Designed for indoor use in construction and heavy-duty maintenance.
  • Meets UL 723 and ASTM E84 standards for flame spread.
Catalog No. 50-275-9574 Supplier Grainger Supplier No. 163K19
3M™ Duct Tape 3939 is a high-performance tape designed for demanding industrial and construction applications. It features a waterproof polyethylene film laminated to a cloth scrim backing with an aggressive rubber adhesive, providing excellent holding power, moisture resistance, and conformability for a variety of tasks including sealing, patching, bundling, and reinforcing.
  • Features a rubber adhesive that adheres aggressively to most surfaces for a tight seal.
  • Polyethylene-coated cloth scrim resists moisture to prolong bond integrity.
  • Offers high conformability and ease of tearing by hand.
  • Low tack roll edges help maintain cleaner surfaces and reduce waste.
  • Suitable for temperatures up to 93°C (200°F).
  • Designed for indoor use in construction and heavy-duty maintenance.
  • Meets UL 723 and ASTM E84 standards for flame spread.