Learn More
Grainger 3M Extra Heavy Duty Duct Tape, Silver, 2 in. x 60 yd., 1 ea.
Catalog No. 502756118
Encompass_Preferred
This extra heavy-duty duct tape features an aggressive rubber adhesive on a polyethylene-coated cloth scrim backing, providing industrial-strength performance for demanding maintenance, repair, and construction tasks. It offers high tack for immediate adhesion to rough or irregular surfaces, excellent abrasion resistance, and moisture protection, making it suitable for HVAC, industrial, and automotive applications.
  • Provides industrial-strength adhesion and 34 lbs./in. tensile strength.
  • Offers up to 60-day clean removal from opaque, non-glass indoor surfaces.
  • Resists water, wear, and abrasion through a durable polyethylene-coated cloth backing.
  • Sticks immediately and holds reliably on rough surfaces.
  • Withstands temperatures up to 200°F.
  • Meets UL 723 flammability, HUD, and BOCA codes.
  • Features easy hand tearability for efficient application.
Catalog No. 50-275-6118 Supplier Grainger Supplier No. 15F769
This extra heavy-duty duct tape features an aggressive rubber adhesive on a polyethylene-coated cloth scrim backing, providing industrial-strength performance for demanding maintenance, repair, and construction tasks. It offers high tack for immediate adhesion to rough or irregular surfaces, excellent abrasion resistance, and moisture protection, making it suitable for HVAC, industrial, and automotive applications.
  • Provides industrial-strength adhesion and 34 lbs./in. tensile strength.
  • Offers up to 60-day clean removal from opaque, non-glass indoor surfaces.
  • Resists water, wear, and abrasion through a durable polyethylene-coated cloth backing.
  • Sticks immediately and holds reliably on rough surfaces.
  • Withstands temperatures up to 200°F.
  • Meets UL 723 flammability, HUD, and BOCA codes.
  • Features easy hand tearability for efficient application.